Please login to your online account to display your discounted pricing

anti-APE, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against KLH-conjugated synthetic peptides corresponding to amino acids 36-52 and 216-232 of human APE.
Antigen APE
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056359T

 
novus biologicals canada
NB10056359SS
Unlabeled 0.025mg Each for $139.00

NB10056359

 
novus biologicals canada
NB10056359
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against KLH-conjugated synthetic peptides corresponding to amino acids 36-52 and 216-232 of human APE.
Antigen APE
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity In HeLa, a 37 kDa band is observed.
Gene Accession No. NP_001632
Gene ID (Entrez) 328
Formulation PBS
Concentration 0.5mg/mL