Please login to your online account to display your discounted pricing

anti-APE, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056359SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056359T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 328
Formulation PBS
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Format Purified
Cross Reactivity Human
Gene Accession No. NP_001632
Test Specificity In HeLa, a 37 kDa band is observed.
Concentration 0.5mg/mL
Host Species Rabbit
Quantity 0.025mg
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Immunogen This antibody was developed against KLH-conjugated synthetic peptides corresponding to amino acids 36-52 and 216-232 of human APE.
Antigen APE