Please login to your online account to display your discounted pricing

anti-APE, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056359

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056359

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_001632
Test Specificity In HeLa, a 37 kDa band is observed.
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Purified
Gene ID (Entrez) 328
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Antigen APE
Formulation PBS
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen This antibody was developed against KLH-conjugated synthetic peptides corresponding to amino acids 36-52 and 216-232 of human APE.
Cross Reactivity Human
Quantity 0.1mg
Regulatory Status RUO
Host Species Rabbit