Please login to your online account to display your discounted pricing

anti-APAF-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056072

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056072

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation PBS
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Conjugate Unlabeled
Gene ID (Entrez) 317
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Apaf 1 .
Antigen APAF-1
Quantity 0.05mL
Immunogen A synthetic peptide corresponding to amino acids 264-282 (GPKYVVPVESSLGKEKGLE) of human Apaf 1 was used as immunogen; GenBank no. NP_001151.1.
Host Species Rabbit
Regulatory Status RUO
Format Antiserum