Please login to your online account to display your discounted pricing

anti-APAF-1, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 451-468 (TQFQRYHQPHTLSPDQED) of human Apaf1 was used as immunogen; GenBank no. NP_001151.1.
Antigen APAF-1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056073

 
novus biologicals canada
NB10056073
Unlabeled 0.05mL Each for $222.95

NB10056073T

 
novus biologicals canada
NB10056073SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 451-468 (TQFQRYHQPHTLSPDQED) of human Apaf1 was used as immunogen; GenBank no. NP_001151.1.
Antigen APAF-1
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Apaf 1 .
Gene ID (Entrez) 317
Formulation PBS