Please login to your online account to display your discounted pricing

anti-APAF-1, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 264-282 (GPKYVVPVESSLGKEKGLE) of human Apaf 1 was used as immunogen; GenBank no. NP_001151.1.
Antigen APAF-1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056072T

 
novus biologicals canada
NB10056072SS
Unlabeled 0.025mL Each for $144.63

NB10056072

 
novus biologicals canada
NB10056072
Unlabeled 0.05mL Each for $222.95
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 264-282 (GPKYVVPVESSLGKEKGLE) of human Apaf 1 was used as immunogen; GenBank no. NP_001151.1.
Antigen APAF-1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Apaf 1 .
Gene ID (Entrez) 317
Formulation PBS