Please login to your online account to display your discounted pricing

anti-APAF-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056073SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056073T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Formulation PBS
Format Antiserum
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Apaf 1 .
Primary or Secondary Primary
Gene ID (Entrez) 317
Conjugate Unlabeled
Regulatory Status RUO
Cross Reactivity Human
Quantity 0.025mL
Immunogen A synthetic peptide corresponding to amino acids 451-468 (TQFQRYHQPHTLSPDQED) of human Apaf1 was used as immunogen; GenBank no. NP_001151.1.
Antigen APAF-1
Host Species Rabbit