Please login to your online account to display your discounted pricing

anti-AP3D1, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 530-580 of human AP3 Delta was used as the immunogen for the antibody.
Antigen AP3D1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227158

 
novus biologicals canada
NBP227158
Unlabeled 0.1mg Each for $257.41

NBP227158SS

 
novus biologicals canada
NBP227158SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 530-580 of human AP3 Delta was used as the immunogen for the antibody.
Antigen AP3D1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The AP3 amino acid sequence used as immunogen for this antibody is 100% homologous in human, chimpanzee, orangutan, new world monkey, rhesus monkey, and 63% homologous in panda, Carolina anole, opossum, and 58% homologous in taipan, zebra finch, horse, dog, pig and 53% homologous in cow, rat, mouse.
Gene ID (Entrez) 8943
Formulation 100μg antibody antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL