Please login to your online account to display your discounted pricing

anti-Anthrax LF, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056667

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056667

  • / Each
 

Description & Specifications

Specifications

Quantity 0.2mL
Monoclonal or Polyclonal Polyclonal
Test Specificity Anthrax LF .
Formulation PBS containing 0.2% gelatin
Format Antiserum
Antigen Anthrax LF
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 290-306 of the Anthrax LF protein (approx. 90 kD).
Conjugate Unlabeled
Regulatory Status RUO
Applications Western Blotting
Primary or Secondary Primary
Cross Reactivity Human
Host Species Rabbit