Please login to your online account to display your discounted pricing

anti-Anthrax LF, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056666SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056666T

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Format Antiserum
Conjugate Unlabeled
Formulation PBS containing 0.2% gelatin
Monoclonal or Polyclonal Polyclonal
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 47-65 of the Anthrax LF protein (approx. 90 kD).
Regulatory Status RUO
Applications Western Blotting
Host Species Rabbit
Cross Reactivity Human
Quantity 0.05mL
Antigen Anthrax LF