Please login to your online account to display your discounted pricing

anti-Anthrax LF, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056666

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056666

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.2% gelatin
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 47-65 of the Anthrax LF protein (approx. 90 kD).
Regulatory Status RUO
Applications Western Blotting
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Antigen Anthrax LF
Quantity 0.2mL
Conjugate Unlabeled
Primary or Secondary Primary
Format Antiserum
Cross Reactivity Human