Please login to your online account to display your discounted pricing

anti-Anthrax LF, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 779-792 of the Anthrax LF protein (approx. 90 kD).
Antigen Anthrax LF
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056668

 
novus biologicals canada
NB10056668
Unlabeled 0.2mL Each for $255.80

NB10056668T

 
novus biologicals canada
NB10056668SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 779-792 of the Anthrax LF protein (approx. 90 kD).
Antigen Anthrax LF
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Formulation PBS containing 0.2% gelatin