Please login to your online account to display your discounted pricing

anti-Anthrax LF, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 290-306 of the Anthrax LF protein (approx. 90 kD).
Antigen Anthrax LF
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056667

 
novus biologicals canada
NB10056667
Unlabeled 0.2mL Each for $257.41

NB10056667T

 
novus biologicals canada
NB10056667SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 290-306 of the Anthrax LF protein (approx. 90 kD).
Antigen Anthrax LF
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Anthrax LF .
Formulation PBS containing 0.2% gelatin