Please login to your online account to display your discounted pricing

anti-Anthrax LF, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056668SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056668T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Antigen Anthrax LF
Formulation PBS containing 0.2% gelatin
Primary or Secondary Primary
Regulatory Status RUO
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 779-792 of the Anthrax LF protein (approx. 90 kD).
Quantity 0.05mL
Format Antiserum
Cross Reactivity Human
Applications Western Blotting
Host Species Rabbit