Please login to your online account to display your discounted pricing

anti-Anthrax LF, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056668

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056668

  • / Each
 

Description & Specifications

Specifications

Quantity 0.2mL
Primary or Secondary Primary
Formulation PBS containing 0.2% gelatin
Conjugate Unlabeled
Host Species Rabbit
Cross Reactivity Human
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 779-792 of the Anthrax LF protein (approx. 90 kD).
Antigen Anthrax LF
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Format Antiserum