Please login to your online account to display your discounted pricing

anti-Anthrax LF, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056667SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056667T

  • / Each
 

Description & Specifications

Specifications

Antigen Anthrax LF
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Antisera
Formulation PBS containing 0.2% gelatin
Host Species Rabbit
Immunogen The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 290-306 of the Anthrax LF protein (approx. 90 kD).
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity Anthrax LF .
Target Species Human