Please login to your online account to display your discounted pricing

anti-Angiopoietin-like Protein 4/ANGPTL4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP219016SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP219016SS

  • / Each
 

Description & Specifications

Specifications

Antigen Angiopoietin-like Protein 4/ANGPTL4
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS
Gene Accession No. Q9BY76
Host Species Rabbit
Immunogen Synthetic peptide made to an internal sequence of human ANGPTL4 (within residues 150-250) [UniProt Q9BY76].
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 51129
Target Species Human
Target Species Murine