Please login to your online account to display your discounted pricing

anti-Angiopoietin-like Protein 4/ANGPTL4, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal sequence of human ANGPTL4 (within residues 150-250) [UniProt Q9BY76].
Antigen Angiopoietin-like Protein 4/ANGPTL4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP219016

 
novus biologicals canada
NBP219016
Unlabeled 0.1mL Each for $329.00

NBP219016SS

 
novus biologicals canada
NBP219016SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal sequence of human ANGPTL4 (within residues 150-250) [UniProt Q9BY76].
Antigen Angiopoietin-like Protein 4/ANGPTL4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. Q9BY76
Gene ID (Entrez) 51129
Formulation PBS
Concentration 1.0mg/mL