missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Amyloid Precursor Protein Mouse anti-Human, Rat, Clone: 1.D5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB349
Description
This Anti-Amyloid Precursor Protein Antibody, A4(695), clone 1.D5 is validated for use in western blotting for the detection of Amyloid Precursor Protein.Specifications
Amyloid Precursor Protein | |
Monoclonal | |
Unconjugated | |
NM_000484.2; NM_201413.1; NM_201414.1 | |
Mouse | |
100 μg | |
Human, Rat | |
Purified |
Western Blot | |
1.D5 | |
Liquid in 0.02M Phosphate buffer, 0.25M NaCl, pH 7.6 with 0.1% sodium azide. | |
APP, PN-II, AD1, ABETA, CTFgamma, APPI, ABPP, A4, CVAP, PN2, PreA4, AAA | |
Synthetic peptide Alz 90 [amino acids 511-608 of amyloid precursor protein pre A4(695)]. | |
Primary | |
−20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
For Research Use Only