Please login to your online account to display your discounted pricing

anti-Amyloid beta A4 Polyclonal, Thermo Scientific™

Amyloid beta A4 Polyclonal Ab OSA00049W from Thermo Scientific™ specifically detects Amyloid beta A4 in Hu, Ms and Rt samples, and it is validated for IHC and WB

$448.05 - $448.05

Specifications

Antigen Amyloid beta A4
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIOSA00049W

 
pierce biotechnology inc
OSA00049W
Unlabeled 100μL Each for $448.05

PIOSA00050G

 
pierce biotechnology inc
OSA00050G
Unlabeled 500μg Each for $448.05
Description & Specifications

Specifications

Antigen Amyloid beta A4
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Host Species Rabbit
Immunogen A synthetic peptide from aa region 400-450 of human APP conjugated to an immunogenic carrier protein
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

Amyloid beta A4 Polyclonal Ab OSA00049W from Thermo Scientific™ specifically detects Amyloid beta A4 in Hu, Ms and Rt samples, and it is validated for IHC and WB


Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Immunogen: A synthetic peptide from aa region 400-450 of human APP conjugated to an immunogenic carrier protein was used as the antigen
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Alzheimer disease amyloid A4 protein homolog, ABPP, APP, Amyloidogenic glycoprotein, AG, Amyloid beta A4 protein, amyloid beta (A4) precursor protein, AAA, AD1, PN2, ABPP, APPI, CVAP, ABETA, CTF gamma, Peptidase nexin-II, Alzheimer disease
Uniprot Accession: P05067, P08592, P12023
Gene Symbol: APP
Entrez GeneID: 11820, 351, 54226
OSA00049W detects Amyloid beta A4 in Human, Mouse and Rat samples. OSA00049W has been successfully used in Immunohistochemistry and Western Blot procedures. OSA00049W The immunogen is A synthetic peptide from aa region 400-450 of human APP conjugated to an immunogenic carrier protein was used as the antigen.