Please login to your online account to display your discounted pricing

anti-Amyloid beta A4 Polyclonal, Thermo Scientific™

Amyloid beta A4 Polyclonal antibody PA1-18317 from Thermo Scientific™ specifically detects Amyloid beta A4 in Rat samples, and it is validated for Immunohistochemistry

$506.05 - $506.05

Specifications

Antigen Amyloid beta A4
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen C-H(388) A H F Q K A K E R L E A K H R E R(405)
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-571

 
pierce biotechnology inc
PA118318
Unlabeled 100μL Each for $506.05

01-670-570

 
pierce biotechnology inc
PA118317
Unlabeled 100μL Each for $506.05
Description & Specifications

Specifications

Antigen Amyloid beta A4
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen C-H(388) A H F Q K A K E R L E A K H R E R(405)
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Amyloid beta A4 Polyclonal antibody PA1-18317 from Thermo Scientific™ specifically detects Amyloid beta A4 in Rat samples, and it is validated for Immunohistochemistry

APP functions as a cell surface receptor and performs physiological functions on the surface of neurons relevant to neurite growth, neuronal adhesion and axonogenesis. The APP protein is involved in cell mobility and transcription regulation through protein-protein interactions. APP can promote transcription activation through binding to APBB1/Tip60 and inhibit Notch signaling through interaction with Numb. APP couples to apoptosis-inducing pathways such as those mediated by G(O) and JIP. Inhibits G(o) alpha ATPase activity. Acts as a kinesin I membrane receptor, mediating the axonal transport of beta-secretase and presenilin 1.
Host Species: Rabbit
Cross Reactivity: Rat
Isotype: IgG
Immunogen: Synthetic peptides corresponding to residuess C-E(145) T H L H W H T V A K E T(157), C-H(388) A H F Q K A K E R L E A K H R E R(405), and C-K(724) K K
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: APP, ABPP, Alzheimer disease amyloid protein, Cerebral vascular amyloid peptide, CVAP; Protease nexin-II, PN-II, APPI, PreA4, A4, AD1
Uniprot Accession: P08592
Gene Symbol: App
Entrez GeneID: 54226
PA1-18317 detects Amyloid beta A4 in Rat samples. PA1-18317 has been successfully used in Immunohistochemistry procedures. PA1-18317 The immunogen is Synthetic peptides corresponding to residuess C-E(145) T H L H W H T V A K E T(157), C-H(388) A H F Q K A K E R L E A K H R E R(405), and C-K(724) K K.