Please login to your online account to display your discounted pricing

anti-Amyloid beta A4 Polyclonal, Thermo Scientific™


Amyloid beta A4 Polyclonal Ab OSA00049W from Thermo Scientific™ specifically detects Amyloid beta A4 in Hu, Ms and Rt samples, and it is validated for IHC and WB

Manufacturer: pierce biotechnology inc  OSA00050G

 View more versions of this product

Catalog No. PIOSA00050G

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Western Blot
Formulation 500μg purified antibody (1mg/mL) and no preservative
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene Accession No. P05067, P08592, P12023
Gene Alias Alzheimer disease amyloid A4 protein homolog, ABPP, APP, Amyloidogenic glycoprotein, AG, Amyloid beta A4 protein, amyloid beta (A4) precursor protein, AAA, AD1, PN2, ABPP, APPI, CVAP, ABETA, CTF gamma, Peptidase nexin-II, Alzheimer disease
Gene Symbols APP
Species Reactivity Human, Mouse, Rat
Storage Requirements avoiding freeze/thaw cycles.
Gene ID (Entrez) 11820, 351, 54226
Host Species Rabbit
Immunogen A synthetic peptide from aa region 400-450 of human APP conjugated to an immunogenic carrier protein
Isotype IgG
Quantity 500μg
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Antigen Amyloid beta A4
Format Purified

Amyloid beta A4 Polyclonal Ab OSA00049W from Thermo Scientific™ specifically detects Amyloid beta A4 in Hu, Ms and Rt samples, and it is validated for IHC and WB


Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: A synthetic peptide from aa region 400-450 of human APP conjugated to an immunogenic carrier protein
Formulation: 500μg purified antibody (1mg/mL) and no preservative
Synonyms: Alzheimer disease amyloid A4 protein homolog, ABPP, APP, Amyloidogenic glycoprotein, AG, Amyloid beta A4 protein, amyloid beta (A4) precursor protein, AAA, AD1, PN2, ABPP, APPI, CVAP, ABETA, CTF gamma, Peptidase nexin-II, Alzheimer disease
Uniprot Accession: P05067, P08592, P12023
Entrez GeneID: 11820, 351, 54226
Gene Symbol: APP
Reconstitute with 500 ul of distilled water.