Please login to your online account to display your discounted pricing

anti-Amyloid beta A4 Polyclonal, Thermo Scientific™


Amyloid beta A4 Polyclonal Ab OSA00049W from Thermo Scientific™ specifically detects Amyloid beta A4 in Hu, Ms and Rt samples, and it is validated for IHC and WB

Manufacturer: pierce biotechnology inc  OSA00049W

 View more versions of this product

Catalog No. PIOSA00049W

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL unpurified antibody in whole serum and no preservative
Applications Immunohistochemistry
Applications Western Blot
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Gene Accession No. P05067, P08592, P12023
Gene Symbols APP
Storage Requirements avoiding freeze/thaw cycles.
Gene ID (Entrez) 11820, 351, 54226
Gene Alias Alzheimer disease amyloid A4 protein homolog, ABPP, APP, Amyloidogenic glycoprotein, AG, Amyloid beta A4 protein, amyloid beta (A4) precursor protein, AAA, AD1, PN2, ABPP, APPI, CVAP, ABETA, CTF gamma, Peptidase nexin-II, Alzheimer disease
Species Reactivity Human, Mouse, Rat
Antigen Amyloid beta A4
Host Species Rabbit
Immunogen A synthetic peptide from aa region 400-450 of human APP conjugated to an immunogenic carrier protein was used as the antigen
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Quantity 100μL

Amyloid beta A4 Polyclonal Ab OSA00049W from Thermo Scientific™ specifically detects Amyloid beta A4 in Hu, Ms and Rt samples, and it is validated for IHC and WB


Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Immunogen: A synthetic peptide from aa region 400-450 of human APP conjugated to an immunogenic carrier protein was used as the antigen
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Alzheimer disease amyloid A4 protein homolog, ABPP, APP, Amyloidogenic glycoprotein, AG, Amyloid beta A4 protein, amyloid beta (A4) precursor protein, AAA, AD1, PN2, ABPP, APPI, CVAP, ABETA, CTF gamma, Peptidase nexin-II, Alzheimer disease
Uniprot Accession: P05067, P08592, P12023
Entrez GeneID: 11820, 351, 54226
Gene Symbol: APP
Reconstitute with 100 ul of distilled water.