Please login to your online account to display your discounted pricing

anti-AMPD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224508

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224508

  • / Each
 

Description & Specifications

Specifications

Antigen AMPD1
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Affinity Purified
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Host Species Rabbit
Immunogen A portion of amino acids 500-550 of human AMPDA1 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 270
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b), horse, mouse, cow, monkeyand chimpanzee, 93% homologous in chicken and 86% homologous inzebrafish. The approximate 70 kD seen in western blot is blocked by the immunizing peptide.
Target Species Human
Target Species Primate