Please login to your online account to display your discounted pricing

anti-AMPD1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 500-550 of human AMPDA1 was used as the immunogen.
Antigen AMPD1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224508SS

 
novus biologicals canada
NBP224508SS
Unlabeled 0.025mg Each for $109.00

NBP224508

 
novus biologicals canada
NBP224508
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 500-550 of human AMPDA1 was used as the immunogen.
Antigen AMPD1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b), horse, mouse, cow, monkeyand chimpanzee, 93% homologous in chicken and 86% homologous inzebrafish. The approximate 70 kD seen in western blot is blocked by the immunizing peptide.
Gene ID (Entrez) 270
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL