Please login to your online account to display your discounted pricing

anti-AMPD1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224508SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224508SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Gene ID (Entrez) 270
Concentration 0.5mg/mL
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_b), horse, mouse, cow, monkeyand chimpanzee, 93% homologous in chicken and 86% homologous inzebrafish. The approximate 70 kD seen in western blot is blocked by the immunizing peptide.
Conjugate Unlabeled
Quantity 0.025mg
Cross Reactivity Human
Cross Reactivity Primate
Immunogen A portion of amino acids 500-550 of human AMPDA1 was used as the immunogen.
Antigen AMPD1
Host Species Rabbit
Format Affinity Purified