Please login to your online account to display your discounted pricing

anti-ALPPL2, Polyclonal, Novus Biologicals™

$397.00 - $397.00

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the N Terminus Region of the target protein.
Antigen ALPPL2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

31-190-002

 
novus biologicals canada
31190002
Unlabeled 0.05mg Each for $397.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody is specific for the N Terminus Region of the target protein.
Antigen ALPPL2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity This product is specific for Human ALPPL2.
Gene Accession No. P10696
Gene ID (Entrez) 251
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0