Please login to your online account to display your discounted pricing

anti-AlphaB Crystallin/CRYAB, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the human Crystallin AB protein (within residues 100-175). [Swiss-Prot P02511]
Antigen AlphaB Crystallin/CRYAB
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002520

 
novus biologicals canada
NB1002520
Unlabeled 0.1mL Each for $307.00

NB1002520SS

 
novus biologicals canada
NB1002520SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the human Crystallin AB protein (within residues 100-175). [Swiss-Prot P02511]
Antigen AlphaB Crystallin/CRYAB
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Human. Predicted to react with rat, sheep, rabbit, monkey, hamster, cow, and pig based on 100% sequence homology. Immunogen sequence has 92% homology to mouse.
Gene Accession No. P02511
Gene ID (Entrez) 1410
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 0.9mg/mL