Please login to your online account to display your discounted pricing

anti-AlphaB Crystallin/CRYAB, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002520SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002520SS

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. P02511
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Conjugate Unlabeled
Primary or Secondary Primary
Test Specificity Human. Predicted to react with rat, sheep, rabbit, monkey, hamster, cow, and pig based on 100% sequence homology. Immunogen sequence has 92% homology to mouse.
Immunogen A synthetic peptide made to a C-terminal region of the human Crystallin AB protein (within residues 100-175). [Swiss-Prot P02511]
Gene ID (Entrez) 1410
Concentration 0.9mg/mL
Antigen AlphaB Crystallin/CRYAB
Cross Reactivity Human
Host Species Rabbit
Format Affinity Purified
Quantity 0.025mL
Applications Western Blotting