Please login to your online account to display your discounted pricing

anti-AlphaB Crystallin/CRYAB, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002520

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002520

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-citrate/phosphate, pH 7-8
Gene ID (Entrez) 1410
Test Specificity Human. Predicted to react with rat, sheep, rabbit, monkey, hamster, cow, and pig based on 100% sequence homology. Immunogen sequence has 92% homology to mouse.
Concentration 0.9mg/mL
Immunogen A synthetic peptide made to a C-terminal region of the human Crystallin AB protein (within residues 100-175). [Swiss-Prot P02511]
Primary or Secondary Primary
Conjugate Unlabeled
Gene Accession No. P02511
Quantity 0.1mL
Antigen AlphaB Crystallin/CRYAB
Applications Western Blotting
Format Affinity Purified
Cross Reactivity Human
Host Species Rabbit
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal