Please login to your online account to display your discounted pricing

anti-alpha-Methylacyl-CoA Racemase/AMACR, Polyclonal, Novus Biologicals™

$321.00 - $429.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide from human AMACR protein
Antigen alpha-Methylacyl-CoA Racemase/AMACR
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232916

 
novus biologicals canada
NBP232916
Unlabeled 0.5mL Each for $321.00

NBP2329161M

 
novus biologicals canada
NBP2329161ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A synthetic peptide from human AMACR protein
Antigen alpha-Methylacyl-CoA Racemase/AMACR
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. Q9UHK6
Gene ID (Entrez) 23600
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL