Please login to your online account to display your discounted pricing

anti-alpha-Methylacyl-CoA Racemase/AMACR, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329161ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329161M

  • / Each
 

Description & Specifications

Specifications

Isotype IgG
Gene ID (Entrez) 23600
Gene Accession No. Q9UHK6
Conjugate Unlabeled
Concentration 0.2mg/mL
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Host Species Rabbit
Cross Reactivity Human
Immunogen A synthetic peptide from human AMACR protein
Antigen alpha-Methylacyl-CoA Racemase/AMACR
Quantity 1mL
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Format Purified