Please login to your online account to display your discounted pricing

anti-alpha-Methylacyl-CoA Racemase/AMACR, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232916

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232916

  • / Each
 

Description & Specifications

Specifications

Antigen alpha-Methylacyl-CoA Racemase/AMACR
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q9UHK6
Host Species Rabbit
Immunogen A synthetic peptide from human AMACR protein
Isotype IgG
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 23600