Please login to your online account to display your discounted pricing

anti-Alpha Fodrin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP192696SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP192696SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q13813
Gene ID (Entrez) 6709
Conjugate Unlabeled
Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Antigen Alpha Fodrin
Format Antiserum
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen 8th and 9th spectrin repeats of human Alpha Fodrin (amino acids 676-1043). [UniProt No. Q13813]
Primary or Secondary Primary
Host Species Rabbit