Please login to your online account to display your discounted pricing

anti-Alpha Fodrin, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen 8th and 9th spectrin repeats of human Alpha Fodrin (amino acids 676-1043). [UniProt No. Q13813]
Antigen Alpha Fodrin
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP192696

 
novus biologicals canada
NBP192696
Unlabeled 0.1mL Each for $309.05

NBP192696SS

 
novus biologicals canada
NBP192696SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen 8th and 9th spectrin repeats of human Alpha Fodrin (amino acids 676-1043). [UniProt No. Q13813]
Antigen Alpha Fodrin
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. Q13813
Gene ID (Entrez) 6709