Please login to your online account to display your discounted pricing

anti-alpha-Fetoprotein/AFP, Polyclonal, Novus Biologicals™

$110.17 - $367.12

Specifications

Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid peptide near the amino terminus of human Alpha 1 Fetoprotein (NP_001125).
Antigen alpha-Fetoprotein/AFP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP176275

 
novus biologicals canada
NBP176275
Unlabeled 0.1mg Each for $367.12

NBP176275SS

 
novus biologicals canada
NBP176275SS
Unlabeled 0.02mg Each for $110.17
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid peptide near the amino terminus of human Alpha 1 Fetoprotein (NP_001125).
Antigen alpha-Fetoprotein/AFP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. P02771
Gene ID (Entrez) 174
Formulation PBS
Concentration 1mg/mL