Please login to your online account to display your discounted pricing

anti-alpha-Fetoprotein/AFP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP176275SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP176275SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P02771
Immunogen Antibody was raised against a 14 amino acid peptide near the amino terminus of human Alpha 1 Fetoprotein (NP_001125).
Conjugate Unlabeled
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene ID (Entrez) 174
Formulation PBS
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Quantity 0.02mg
Host Species Rabbit
Antigen alpha-Fetoprotein/AFP
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified