Please login to your online account to display your discounted pricing

anti-Aldo-keto Reductase 1B10/AKR1B10, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP144998SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP144998SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. O60218
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Regulatory Status RUO
Test Specificity Reacts with human. Does not cross react with mouse, rat or rabbit.
Concentration 1mg/mL
Formulation 0.1mM Tris-HCl, 100mM sodium chloride and 50% glycerol. (No preservative).
Host Species Rabbit
Gene ID (Entrez) 57016
Cross Reactivity Human
Antigen Aldo-keto Reductase 1B10/AKR1B10
Primary or Secondary Primary
Immunogen A highly specific 15 amino acid portion of human AKR1B10 (within amino acids 100-150). [UniProt No. O60218]
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Quantity 0.025mL