Please login to your online account to display your discounted pricing

anti-Aldo-keto Reductase 1B10/AKR1B10, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP144998

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP144998

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene Accession No. O60218
Test Specificity Reacts with human. Does not cross react with mouse, rat or rabbit.
Gene ID (Entrez) 57016
Formulation 0.1mM Tris-HCl, 100mM sodium chloride and 50% glycerol. (No preservative).
Concentration 1mg/mL
Quantity 0.1mL
Immunogen A highly specific 15 amino acid portion of human AKR1B10 (within amino acids 100-150). [UniProt No. O60218]
Regulatory Status RUO
Conjugate Unlabeled
Antigen Aldo-keto Reductase 1B10/AKR1B10
Cross Reactivity Human
Format Affinity Purified
Host Species Rabbit