Please login to your online account to display your discounted pricing

anti-Aldo-keto Reductase 1B10/AKR1B10, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A highly specific 15 amino acid portion of human AKR1B10 (within amino acids 100-150). [UniProt No. O60218]
Antigen Aldo-keto Reductase 1B10/AKR1B10
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP144998

 
novus biologicals canada
NBP144998
Unlabeled 0.1mL Each for $329.00

NBP144998SS

 
novus biologicals canada
NBP144998SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A highly specific 15 amino acid portion of human AKR1B10 (within amino acids 100-150). [UniProt No. O60218]
Antigen Aldo-keto Reductase 1B10/AKR1B10
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Reacts with human. Does not cross react with mouse, rat or rabbit.
Gene Accession No. O60218
Gene ID (Entrez) 57016
Formulation 0.1mM Tris-HCl, 100mM sodium chloride and 50% glycerol. (No preservative).
Concentration 1mg/mL