Please login to your online account to display your discounted pricing

anti-Aldo-keto Reductase 1B10/AKR1B10, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-200 of human AKR1B10 protein were used as the immunogen.
Antigen Aldo-keto Reductase 1B10/AKR1B10
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056423T

 
novus biologicals canada
NB10056423SS
Unlabeled 0.025mg Each for $139.00

NB10056423

 
novus biologicals canada
NB10056423
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-200 of human AKR1B10 protein were used as the immunogen.
Antigen Aldo-keto Reductase 1B10/AKR1B10
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The NCBI accession number for AKR1B10 is EAL24068. Cross reacts with Mouse, Human and Rat. Predicted to react with Chimpanzee.
Gene Accession No. C9JRZ8
Gene ID (Entrez) 57016
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL