Please login to your online account to display your discounted pricing

anti-Aldo-keto Reductase 1B10/AKR1B10, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056423

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056423

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation PBS containing 0.2% gelatin
Conjugate Unlabeled
Test Specificity The NCBI accession number for AKR1B10 is EAL24068. Cross reacts with Mouse, Human and Rat. Predicted to react with Chimpanzee.
Gene Accession No. C9JRZ8
Gene ID (Entrez) 57016
Format Purified
Applications Western Blotting
Regulatory Status RUO
Quantity 0.1mg
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Primary or Secondary Primary
Immunogen A portion of amino acids 100-200 of human AKR1B10 protein were used as the immunogen.
Antigen Aldo-keto Reductase 1B10/AKR1B10