Please login to your online account to display your discounted pricing

anti-ALDH1L1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225143SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225143SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10840
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Rat. Other species have not been tested.
Quantity 0.025mL
Antigen ALDH1L1
Immunogen Full length purified ALDH1L1
Cross Reactivity Rat
Format Antiserum
Regulatory Status RUO