Please login to your online account to display your discounted pricing

anti-ALDH1L1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225143

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225143

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Gene ID (Entrez) 10840
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Rat. Other species have not been tested.
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mL
Host Species Rabbit
Format Antiserum
Primary or Secondary Primary
Cross Reactivity Rat
Antigen ALDH1L1
Immunogen Full length purified ALDH1L1