Please login to your online account to display your discounted pricing

anti-ALDH1L1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Full length purified ALDH1L1
Antigen ALDH1L1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225143

 
novus biologicals canada
NBP225143
Unlabeled 0.1mL Each for $309.05

NBP225143SS

 
novus biologicals canada
NBP225143SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Full length purified ALDH1L1
Antigen ALDH1L1
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Rat. Other species have not been tested.
Gene ID (Entrez) 10840