Please login to your online account to display your discounted pricing

anti-Akt3, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was raised against a synthetic peptide corresponding to amino acid residues 119-136 of human AKT3.
Antigen Akt3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056345

 
novus biologicals canada
NB10056345
Unlabeled 0.1mg Each for $255.80

NB10056345T

 
novus biologicals canada
NB10056345SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This polyclonal antibody was raised against a synthetic peptide corresponding to amino acid residues 119-136 of human AKT3.
Antigen Akt3
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity A 50 kDa band is observed.
Gene Accession No. Q9Y243
Gene ID (Entrez) 10000
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL