Please login to your online account to display your discounted pricing

anti-AKT2/3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056362

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056362

  • / Each
 

Description & Specifications

Specifications

Antigen AKT2/3
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS
Gene Accession No. NP_001617
Host Species Rabbit
Immunogen Amino acids 319-331 of human Akt2. This sequence is 100% homologous in Akt3.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 208
Target Species Human