Please login to your online account to display your discounted pricing

anti-AKT2/3, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Amino acids 319-331 of human Akt2. This sequence is 100% homologous in Akt3.
Antigen AKT2/3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056362

 
novus biologicals canada
NB10056362
Unlabeled 0.1mg Each for $255.80

NB10056362T

 
novus biologicals canada
NB10056362SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 319-331 of human Akt2. This sequence is 100% homologous in Akt3.
Antigen AKT2/3
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. NP_001617
Gene ID (Entrez) 208
Formulation PBS
Concentration 0.5mg/mL