Please login to your online account to display your discounted pricing

anti-AKT2/3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056362SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056362T

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Primary or Secondary Primary
Gene ID (Entrez) 208
Concentration 0.5mg/mL
Conjugate Unlabeled
Applications Western Blotting
Gene Accession No. NP_001617
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mg
Cross Reactivity Human
Immunogen Amino acids 319-331 of human Akt2. This sequence is 100% homologous in Akt3.
Format Purified
Host Species Rabbit
Regulatory Status RUO
Antigen AKT2/3