Please login to your online account to display your discounted pricing

anti-Akt1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056631

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056631

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_001014432
Formulation PBS and 0.1mg/mL BSA
Concentration 0.1mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Quantity 0.2mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Test Specificity Cross reacts with Human. Predicted to react with Mouse, Frog and Chicken.
Gene ID (Entrez) 207
Format Purified
Immunogen This polyclonal antibody was developed against a synthetic peptide corresponding to amino acid residues 464-477 of human Akt1.
Host Species Rabbit
Regulatory Status RUO
Antigen Akt1
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human